Boc-Glu-OH

Supplier: BACHEM AMERICAS INC

4014187.0025 4014187.0100
A-3620.0025BAEA 284.04 CAD
A-3620.0025BA A-3620.0100BA
Boc-Glu-OH
Boc-Glu-OH

Sequence: Boc-Glu-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₁₀H₁₇NO₆
Storage Temperature: Refrigerator
MDL Number: MFCD00037297
CAS Number: 2419-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR