N-(tert-Butoxycarbonyl)-L-aspartic acid

Supplier: BACHEM AMERICAS INC

4016413.0025 4016413.0100
A-3615.0025BAEA 183.09 CAD
A-3615.0025BA A-3615.0100BA
N-(tert-Butoxycarbonyl)-L-aspartic acid
N-(tert-Butoxycarbonyl)-L-aspartic acid

Sequence: Boc-Asp-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₉H₁₅NO₆
Storage Temperature: Refrigerator
MDL Number: MFCD00037279
CAS Number: 13726-67-5
EINECS: 237-294-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR