Boc-S-trityl-L-homocysteine

Supplier: BACHEM AMERICAS INC

4025120.0001 4025120.0005
A-3610.0001BAEA 903.62 CAD
A-3610.0001BA A-3610.0005BA
Boc-S-trityl-L-homocysteine
Boc-S-trityl-L-homocysteine

Sequence: Boc-Homocys(Trt)-OH


The cysteine homolog is formed by S-demethylation of methionine. Contrary to Cys, Hcy can be incorporated and derivatized without risk of racemization and β-elimination. Base-catalyzed desulfuration of peptides containing a Cys/Hcy mixed disulfide bridge yields cystathionine (carbacystine) peptides.


Synonym(s): (S)-2-(Boc-amino)-4-tritylsulfanyl-butyric acid

Formula: C₂₈H₃₁NO₄S
Storage Temperature: Freezer
MDL Number: MFCD00672309
CAS Number: 201419-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR