Boc-4-azido-Phe-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-p-azido-Phe-OH

4020249.0001 4020249.0005
A-3570.0001BAEA 748.67 CAD
A-3570.0001BA A-3570.0005BA
Boc-4-azido-Phe-OH
Boc-4-azido-Phe-OH

Sequence: Boc-p-azido-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.
For further azido compounds suitable as photolabels or for 1,3-dipolar cycloaddition / click chemistry please see Q-1995 (3-(4-Azidophenyl)propionic acid), Q-2005 (4-(4-Azidophenyl)butyric acid and Q-2725 (5-Azido-pentanoic acid). Azido-functionalized amino acid derivatives are also on offer: Fmoc-Dab(N₂)-OH (B-4145), Fmoc-Orn(N₂)-OH (B-4140), Fmoc-Lys(N₂)-OH (B-4135), Boc-Orn(N₂)-OH · DCHA (A-4770), and Boc-Lys(N₂)-OH · DCHA (A-4765).

Formula: C₁₄H₁₈N₄O₄
Storage Temperature: Freezer
MDL Number: MFCD00237570
CAS Number: 33173-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR