Boc-Arg(Pbf)-OH

Supplier: BACHEM AMERICAS INC

4019088.0001 4019088.0005 4019088.0025
A-3565.0001BAEA 180.91 CAD
A-3565.0001BA A-3565.0005BA A-3565.0025BA
Boc-Arg(Pbf)-OH
Boc-Arg(Pbf)-OH

Sequence: Boc-Arg(Pbf)-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₂₄H₃₈N₄O₇S
Storage Temperature: Freezer
MDL Number: MFCD00236821
CAS Number: 200124-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR