Boc-S-trityl-L-penicillamine

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-Pen(Trt)-OH

4019781.0001 4019781.0005
A-3550.0001BAEA 608.66 CAD
A-3550.0001BA A-3550.0005BA
Boc-S-trityl-L-penicillamine
Boc-S-trityl-L-penicillamine

Sequence: Boc-Pen(Trt)-OH


Corresponds to β,β-dimethylcysteine or β-mercaptovaline. For an S-protected derivative of deamino-penicillamine please see Q-1610. The rigidity of disulfide bridges can be increased by replacing Cys by Pen. Pen analogs of enkephalins, RGD-peptides, urotensin and further active peptides with improved potency and/or selectivity have been described. Bachem offers a choice of resins preloaded with Fmoc-Pen derivatives, please see the product family 'Preloaded Resins for SPPS and SPOS'.


Synonym(s): Boc-β,β-dimethyl-Cys(Trt)-OH

Formula: C₂₉H₃₃NO₄S
MW: 491.646 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237381
CAS Number: 135592-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR