H-Orn(Boc)-OMe·HCl

Supplier: BACHEM AMERICAS INC

4018999.0001 4018999.0005
A-3490.0001BAEA 284.04 CAD
A-3490.0001BA A-3490.0005BA
H-Orn(Boc)-OMe·HCl
H-Orn(Boc)-OMe·HCl

Sequence: H-Orn(Boc)-OMe · HCl

Formula: C₁₁H₂₃ClN₂O₄
Storage Temperature: Freezer
MDL Number: MFCD00237207
CAS Number: 2480-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR