Boc-Glu-OMe
Supplier: BACHEM AMERICAS INC
Synonyms:
(S)-4-((tert-Butoxycarbonyl)amino)-5-methoxy-5-oxopentanoic acid, Boc-Glu-OMe
4018552.0005
4018552.0025
A-3440.0005BAEA
464.57
CAD
A-3440.0005BA
A-3440.0025BA
Boc-Glu-OMe
Boc-Glu-OMe
Sequence: Boc-Glu-OMe
The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.
Formula:
C₁₁H₁₉NO₆ Storage Temperature: Refrigerator |
MDL Number:
MFCD00076931 CAS Number: 72086-72-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...