Boc-Cys(Fm)-OH

Supplier: BACHEM AMERICAS INC

4018970.0001 4018970.0005
A-3435.0001BAEA 335.58 CAD
A-3435.0001BA A-3435.0005BA
Boc-Cys(Fm)-OH
Boc-Cys(Fm)-OH

Sequence: Boc-Cys(Fm)-OH


Fm can be removed on-resin allowing disulfide bridge formation on solid phase.

Formula: C₂₂H₂₅NO₄S
Storage Temperature: Freezer
MDL Number: MFCD00076921
CAS Number: 84888-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR