Boc-D-Glu(Ochex)-OH
Supplier: BACHEM AMERICAS INC
4018226.0001
4018226.0005
A-3425.0001BAEA
201.47
CAD
A-3425.0001BA
A-3425.0005BA
Boc-D-Glu(Ochex)-OH
Boc-D-Glu(Ochex)-OH
Sequence: Boc-D-Glu(OcHex)-OH
The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.
Formula:
C₁₆H₂₇NO₆ Storage Temperature: Refrigerator |
MDL Number:
MFCD00076929 CAS Number: 133464-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...