Boc-D-ORN-OH

Supplier: BACHEM AMERICAS INC

4019006.0001 4019006.0005
A-3380.0001BAEA 439.41 CAD
A-3380.0001BA A-3380.0005BA
Boc-D-ORN-OH
Boc-D-ORN-OH

Sequence: Boc-D-Orn-OH

Formula: C₁₀H₂₀N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00236869
CAS Number: 159877-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR