Boc-allo-Ile-OH

Supplier: BACHEM AMERICAS INC

4012081.0001 4012081.0005
A-3345.0001BAEA 645.43 CAD
A-3345.0001BA A-3345.0005BA
Boc-allo-Ile-OH
Boc-allo-Ile-OH

Sequence: Boc-allo-Ile-OH

Formula: C₁₁H₂₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD00151905
CAS Number: 35264-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR