Boc-Orn(Fmoc)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Nα-Boc-Nδ-Fmoc-L-ornithine

4017884.0001 4017884.0005 4017884.0025
A-3325.0001BAEA 180.91 CAD
A-3325.0001BA A-3325.0005BA A-3325.0025BA
Boc-Orn(Fmoc)-OH
Boc-Orn(Fmoc)-OH

Sequence: Boc-Orn(Fmoc)-OH

Formula: C₂₅H₃₀N₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00080279
CAS Number: 150828-96-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR