Boc-S-trityl-D-cysteine

Supplier: BACHEM AMERICAS INC

4017511.0001 4017511.0005
A-3265.0001BAEA 211.67 CAD
A-3265.0001BA A-3265.0005BA
Boc-S-trityl-D-cysteine
Boc-S-trityl-D-cysteine

Sequence: Boc-D-Cys(Trt)-OH

Formula: C₂₇H₂₉NO₄S
Storage Temperature: Freezer
MDL Number: MFCD00236839
CAS Number: 87494-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR