Boc-2-Abz-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-Abz-OH

4002711.0005 4002711.0025 4002711.0100
A-3240.0005BAEA 180.91 CAD
A-3240.0005BA A-3240.0025BA A-3240.0100BA
Boc-2-Abz-OH
Boc-2-Abz-OH

Sequence: Boc-Abz-OH
Synonym(s): Boc-anthranilic acid

Formula: C₁₂H₁₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD00151862
CAS Number: 68790-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR