(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid

Supplier: BACHEM AMERICAS INC

Synonyms: 3-Amino-N-Boc-L-alanine, Nalpha-Boc-2,3-diaminopropionic Acid, Boc-Dpr-OH, Boc-Dap-OH, 3-Amino-N-(tert-butoxycarbonyl)-L-alanine

4015642.0001 4015642.0005
A-3220.0001BAEA 206.51 CAD
A-3220.0001BA A-3220.0005BA
(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid
(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid

Sequence: Boc-Dap-OH


Abbreviated Dap, the abbreviation Dpr can be found in the literature as well. See also Albiziine, Nβ-carbamoyl-Dab, F-3670.

Formula: C₈H₁₆N₂O₄
MW: 204.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236843
CAS Number: 73259-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR