(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid
Supplier: BACHEM AMERICAS INC
Synonyms:
Boc-Dab-OH, (S)-4-Amino-2-(Boc-amino)butyric Acid, (S)-4-Amino-2-(tert-butoxycarbonylamino)butyric Acid, (S)-Nalpha-Boc-2,4-diaminobutyric Acid, (S)-4-Amino-2-(Boc-amino)butyric acid (S)-Nα-Boc-2,4-diaminobutyric acid Boc-Dab-OH
4016073.0001
4016073.0005
4016073.0025
A-3215.0001BAEA
211.77
CAD
A-3215.0001BA
A-3215.0005BA
A-3215.0025BA
(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid
(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid
Sequence: Boc-Dab-OH
Formula:
C₉H₁₈N₂O₄ MW: 218.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00236841 CAS Number: 25691-37-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...