Boc-Cit-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-5-ureidopentanoic acid

4000079.0005 4000079.0025
A-3165.0005BAEA 309.81 CAD
A-3165.0005BA A-3165.0025BA
Boc-Cit-OH
Boc-Cit-OH

Sequence: Boc-Cit-OH


Bachem also offers Fmoc-Cit-Wang resin, D-1840.


Synonym(s): Boc-Orn(carbamoyl)-OH

Formula: C₁₁H₂₁N₃O₅
Storage Temperature: Freezer
MDL Number: MFCD00080271
CAS Number: 45234-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR