Boc-Asp-OtBu

Supplier: BACHEM AMERICAS INC

4026472.0001 4026472.0005 4026472.0025
A-3120.0001BAEA 185.37 CAD
A-3120.0001BA A-3120.0005BA A-3120.0025BA
Boc-Asp-OtBu
Boc-Asp-OtBu
Formula: C₁₃H₂₃NO₆
Storage Temperature: Freezer
MDL Number: MFCD00038272
CAS Number: 34582-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR