Boc-β-t-butyl-L-alanine

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-β-tBu-Ala-OH, (S)-2-((tert-Butoxycarbonyl)amino)-4,4-dimethylpentanoic acid, Boc-β-tBuAla-OH

4015631.0001 4015631.0005
A-3110.0001BAEA 672.04 CAD
A-3110.0001BA A-3110.0005BA
Boc-β-t-butyl-L-alanine
Boc-beta-t-butyl-L-alanine

Sequence: Boc-β-tBu-Ala-OH


Besides Ala derivatives, Bachem offers a number of derivatives of β-substituted alanines, amongst them: t-Butylalanine Cyanoalanine Cyclohexylalanine (Cha) Cyclopropylalanine 2-Quinolylalanine β-Halogenated Alanines
γ-Methylleucine, also called neopentylglycine.


Synonym(s): Boc-Neopentylgly-OH#Boc-γ-Me-Leu-OH#Boc-Tba-OH

Formula: C₁₂H₂₃NO₄
MW: 245.319 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00237540
CAS Number: 79777-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR