Boc-Ser-Otbu

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-Ser-OtBu

4001192.0001 4001192.0005
A-3025.0001BAEA 217.06 CAD
A-3025.0001BA A-3025.0005BA
Boc-Ser-Otbu
Boc-Ser-Otbu

Sequence: Boc-Ser-OtBu


Bachem also offers derivatives of phosphoserine and sulfoserine.

Formula: C₁₂H₂₃NO₅
MW: 261.318 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00190832
CAS Number: 7738-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR