4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate
Supplier: BACHEM AMERICAS INC
Synonyms:
4-tert-Butyl N-Boc-L-aspartate, N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-tert-Butyl Ester, N-Boc-L-aspartic Acid 4-tert-Butyl Ester, Boc-Asp(OtBu)-OH
4025872.0001
4025872.0005
A-2950.0001BAEA
185.46
CAD
A-2950.0001BA
A-2950.0005BA
4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate
4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate
Sequence: Boc-Asp(OtBu)-OH
Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.
Formula:
C₁₃H₂₃NO₆ MW: 289.33 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00076912 CAS Number: 1676-90-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...