Boc-Glu(Osu)-Otbu
Supplier: BACHEM AMERICAS INC
4014280.0001
4014280.0005
4014280.0025
A-2910.0001BAEA
180.91
CAD
A-2910.0001BA
A-2910.0005BA
A-2910.0025BA
Boc-Glu(Osu)-Otbu
Boc-Glu(Osu)-Otbu
Sequence: Boc-Glu(OSu)-OtBu
The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.
Formula:
C₁₈H₂₈N₂O₈ Storage Temperature: Freezer |
MDL Number:
MFCD00236848 CAS Number: 81659-82-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...