Boc-Orn-OH

Supplier: BACHEM AMERICAS INC

4014396.0005 4014396.0025
A-2865.0005BAEA 180.91 CAD
A-2865.0005BA A-2865.0025BA
Boc-Orn-OH
Boc-Orn-OH

Sequence: Boc-Orn-OH


Educt for the synthesis of Boc- and Fmoc-δ-azido-norvaline.

Formula: C₁₀H₂₀N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00076970
CAS Number: 21887-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR