Boc-hyp(Bzl)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-Hyp(Bzl)-OH

4014352.0001 4014352.0005
A-2820.0001BAEA 180.91 CAD
A-2820.0001BA A-2820.0005BA A-2820.0025BA
Boc-hyp(Bzl)-OH
Boc-hyp(Bzl)-OH

Sequence: Boc-Hyp(Bzl)-OH


Hyp denotes L-trans-hydroxyproline.

Formula: C₁₇H₂₃NO₅
Storage Temperature: Ambient
MDL Number: MFCD00037325
CAS Number: 54631-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR