5-Cyclohexyl-N-(tert-butoxycarbonyl)-L-glutamate
Supplier: BACHEM AMERICAS INC
Synonyms:
5-Cyclohexyl N-Boc-L-glutamate, Boc-Glu(OcHex)-OH, N-(tert-Butoxycarbonyl)-L-glutamic Acid 5-Cyclohexyl Ester, N-Boc-L-glutamic Acid 5-Cyclohexyl Ester
4030405.0025
4030405.0100
A-2770.0025BAEA
328.36
CAD
A-2770.0025BA
A-2770.0100BA
5-Cyclohexyl-N-(tert-butoxycarbonyl)-L-glutamate
5-Cyclohexyl-N-(tert-butoxycarbonyl)-L-glutamate
Sequence: Boc-Glu(OcHex)-OH
The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.
Formula:
C₁₆H₂₇NO₆ MW: 329.39 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00065570 CAS Number: 73821-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...