Boc-His(3-Me)-OH

Supplier: BACHEM AMERICAS INC

4011125.0250 4011125.1000
A-2565.0250BAEA 317.74 CAD
A-2565.0250BA A-2565.1000BA
Boc-His(3-Me)-OH
Boc-His(3-Me)-OH

Sequence: Boc-His(3-Me)-OH


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Synonym(s): Boc-His(π-Me)-OH

Formula: C₁₂H₁₉N₃O₄
Storage Temperature: Freezer
MDL Number: MFCD00076945
CAS Number: 61070-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR