Boc-D-Trp(For)-OH

Supplier: BACHEM AMERICAS INC

4010099.0001 4010099.0005
A-2500.0001BAEA 227.28 CAD
A-2500.0001BA A-2500.0005BA
Boc-D-Trp(For)-OH
Boc-D-Trp(For)-OH

Sequence: Boc-D-Trp(For)-OH

Formula: C₁₇H₂₀N₂O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00038004
CAS Number: 64905-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR