N-α-Boc-N-β-xanthenyl-L-asparagine

Supplier: BACHEM AMERICAS INC

4007622.0025 4007622.0100
A-2485.0025BAEA 400.8 CAD
A-2485.0025BA A-2485.0100BA
N-α-Boc-N-β-xanthenyl-L-asparagine
N-alpha-Boc-N-beta-xanthenyl-L-asparagine

Sequence: Boc-Asn(Xan)-OH


For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".

Formula: C₂₂H₂₄N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00066157
CAS Number: 65420-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR