Boc-L-valine hydroxysuccinimide ester

Supplier: BACHEM AMERICAS INC

Synonyms: N-Boc-L-valine N-succinimidyl ester, Boc-Val-OSu

4000117.0005 4000117.0025
A-2470.0005BAEA 183.09 CAD
A-2470.0005BA A-2470.0025BA
Boc-L-valine hydroxysuccinimide ester
Boc-L-valine hydroxysuccinimide ester

Sequence: Boc-Val-OSu


Bachem additionally offers Fmoc-[¹⁵N]Val-OH, B-2670.

Formula: C₁₄H₂₂N₂O₆
MW: 314.338 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00037906
CAS Number: 3392-12-9
EINECS: 222-236-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR