N-Boc-L-valine

Supplier: BACHEM AMERICAS INC

4000426.0100 4000426.0500 4000426.1000
A-2455.0100BAEA 227.28 CAD
A-2455.0100BA A-2455.0500BA A-2455.1000BA
N-Boc-L-valine
N-Boc-L-valine

Sequence: Boc-Val-OH


Bachem additionally offers Fmoc-[¹⁵N]Val-OH, B-2670.

Formula: C₁₀H₁₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD00065605
CAS Number: 13734-41-3
EINECS: 237-307-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR