Boc-L-tyrosine methyl ester
Supplier: BACHEM AMERICAS INC
Synonyms:
Boc-Tyr-OMe, N-Boc-L-tyrosine Methyl Ester
4000251.0025
4000251.0100
A-2420.0025BAEA
335.58
CAD
A-2420.0025BA
A-2420.0100BA
Boc-L-tyrosine methyl ester
Boc-L-tyrosine methyl ester
Sequence: Boc-Tyr-OMe
Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.
Educt for Zn(II) and Co(II)-complexing Fréchet-type polyaryl ether dendrons.
Formula:
C₁₅H₂₁NO₅ Storage Temperature: Freezer |
MDL Number:
MFCD00191181 CAS Number: 4326-36-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...