N-Boc-L-tyrosine

Supplier: BACHEM AMERICAS INC

4000198.0025 4000198.0100
A-2405.0025BAEA 185.3 CAD
A-2405.0025BA A-2405.0100BA
N-Boc-L-tyrosine
N-Boc-L-tyrosine

Sequence: Boc-Tyr-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₁₄H₁₉NO₅
Storage Temperature: Ambient
MDL Number: MFCD00037179
CAS Number: 3978-80-1
EINECS: 223-613-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR