N-Boc-S-trityl-L-cysteine

Supplier: BACHEM AMERICAS INC

Synonyms: N-Boc-S-trityl-L-cysteine, N-(tert-Butoxycarbonyl)-S-trityl-L-cysteine, BOC-S-trityl-L-cysteine, (R)-2-((tert-Butoxycarbonyl)amino)-3-(tritylthio)propanoic acid, Boc-Cys(Trt)-OH

4003922.0025 4003922.0100
A-2355.0025BAEA 434.83 CAD
A-2355.0025BA A-2355.0100BA
N-Boc-S-trityl-L-cysteine
N-Boc-S-trityl-L-cysteine

Sequence: Boc-Cys(Trt)-OH

Formula: C₂₇H₂₉NO₄S
MW: 463.592 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038251
CAS Number: 21947-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR