Nɑ-Boc-N-ω-tosyl-L-arginine

Supplier: BACHEM AMERICAS INC

Synonyms: Nɑ-(tert-Butoxycarbonyl)-Nω-(p-toluenesulfonyl)-L-arginine, Boc-Arg(Tos)-OH, Nɑ-(tert-Butoxycarbonyl)-Nω-(p-toluenesulphonyl)-L-arginine

4001496.0025 4001496.0100
A-2315.0025BAEA 276.63 CAD
A-2315.0025BA A-2315.0100BA
Nɑ-Boc-N-ω-tosyl-L-arginine
Nalpha-Boc-N-omega-tosyl-L-arginine

Sequence: Boc-Arg(Tos)-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.


A suitable derivative for solid-phase synthesis. The tosyl group can be removed efficiently by treatment with HF/anisole or TFMSA/TFA/thioanisole.

Formula: C₁₈H₂₈N₄O₆S
MW: 428.50 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00065557
CAS Number: 13836-37-8
EINECS: 237-549-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR