N-Boc-L-threonine

Supplier: BACHEM AMERICAS INC

4000124.0025 4000124.0100
A-2305.0025BAEA 187.69 CAD
A-2305.0025BA A-2305.0100BA
N-Boc-L-threonine
N-Boc-L-threonine

Sequence: Boc-Thr-OH


Bachem also offers phosphothreonine derivatives.


Building block for the incorporation of O-acyl isopeptide bonds during Fmoc-SPPS.

Formula: C₉H₁₇NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD00065946
CAS Number: 2592-18-9
EINECS: 219-987-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR