N-Boc-L-serine
Supplier: BACHEM AMERICAS INC
Synonyms:
(S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxypropanoic acid, Boc-Ser-OH, N-(tert-Butoxycarbonyl)-L-serine, Boc-L-serine, N-(tert-Butoxycarbonyl)-L-serine hydrate, BOC-L-Serine hydrate
Sequence: Boc-Ser-OH
Bachem also offers derivatives of phosphoserine and sulfoserine.
Educt for the synthesis of Boc-Ser β-lactone, a useful synthon for obtaining stereochemically pure β-substituted α-amino acids. Building block for the incorporation of O-acyl isopeptide bonds during Fmoc-SPPS.
Formula:
C₈H₁₅NO₅ Storage Temperature: Refrigerator |
MDL Number:
MFCD00037243 CAS Number: 3262-72-4 EINECS: 221-867-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...