Boc-sarcosine

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-Sar-OH

4001115.0025 4001115.0100
A-2265.0025BAEA 180.91 CAD
A-2265.0025BA A-2265.0100BA
Boc-sarcosine
Boc-sarcosine

Sequence: Boc-Sar-OH


Bachem also offers Fmoc-Sar-Wang resin (D-1805) and Fmoc-Sar-SASRIN resin (-D-1645).


Synonym(s): Boc-N-Me-Gly-OH

Formula: C₈H₁₅NO₄
MW: 189.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037795
CAS Number: 13734-36-6
EINECS: 237-306-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR