BOC-L-phenylalanine-N-hydroxysuccinimide ester

Supplier: BACHEM AMERICAS INC

4001917.0005 4001917.0025
A-2175.0005BAEA 187.69 CAD
A-2175.0005BA A-2175.0025BA
BOC-L-phenylalanine-N-hydroxysuccinimide ester
BOC-L-phenylalanine-N-hydroxysuccinimide ester

Sequence: Boc-Phe-OSu


Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.

Formula: C₁₈H₂₂N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00037911
CAS Number: 3674-06-4
EINECS: 222-939-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR