N-(tert-Butoxycarbonyl)-D-phenylalanine
Supplier: BACHEM AMERICAS INC
4000781.0025
4000781.0100
A-2165.0025BAEA
336.29
CAD
A-2165.0025BA
A-2165.0100BA
N-(tert-Butoxycarbonyl)-D-phenylalanine
N-(tert-Butoxycarbonyl)-D-phenylalanine
Sequence: Boc-D-Phe-OH
Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.
Formula:
C₁₄H₁₉NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00063149 CAS Number: 18942-49-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...