N-(tert-Butoxycarbonyl)-L-phenylalanine

Supplier: BACHEM AMERICAS INC

Synonyms: N-Boc-L-phenylalanine, BOC-L-Phenylalanine, N-α-t-Boc-L-phenylalanine, Boc-Phe-OH

4000663.0100 4000663.0500 4000663.1000
A-2160.0100BAEA 241.82 CAD
A-2160.0100BA A-2160.0500BA A-2160.1000BA
N-(tert-Butoxycarbonyl)-L-phenylalanine
N-(tert-Butoxycarbonyl)-L-phenylalanine

Sequence: Boc-Phe-OH


Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002663
CAS Number: 13734-34-4
EINECS: 237-305-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR