Boc-Arg(NO₂)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Nα-Boc-Nomega-nitro-L-arginine

4001219.0025 4001219.0100
A-2115.0025BAEA 493.53 CAD
A-2115.0025BA A-2115.0100BA
Boc-Arg(NO₂)-OH
Boc-Arg(NO₂)-OH

Sequence: Boc-Arg(NO₂)-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.


The nitro group of the guanidyl residue can be removed by treating the blocked peptide with HF at 0 °C for 40 min.

Formula: C₁₁H₂₁N₅O₆
MW: 319.318 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00065556
CAS Number: 2188-18-3
EINECS: 218-580-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR