Boc-Arg(Mts)-OH cyclohexylammonium salt

Supplier: BACHEM AMERICAS INC

Synonyms: α-Boc-Arg(Nω-Mesitylenesulfonyl)-OH cyclohexylammonium salt, α-Boc-Arg(Nω-Mesitylenesulphonyl)-OH cyclohexylammonium salt, Boc-Arg(Mts)-OH·CHA

4010095.0005 4010095.0025
A-2110.0005BAEA 309.81 CAD
A-2110.0005BA A-2110.0025BA
Boc-Arg(Mts)-OH cyclohexylammonium salt
Boc-Arg(Mts)-OH cyclohexylammonium salt

Sequence: Boc-Arg(Mts)-OH · CHA


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.


Mts is quantitatively cleaved by treatment with HF, TFMSA, or methanesulfonic acid.

Formula: C₂₀H₃₂N₄O₆S·C₆H₁₃N
MW: 555.73 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00057790
CAS Number: 68262-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR