Boc-Arg(Mtr)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Nalpha-Boc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulphonyl)-L-arginine, Nalpha-Boc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine

4007638.0005 4007638.0025
A-2105.0005BAEA 464.74 CAD
A-2105.0005BA A-2105.0025BA
Boc-Arg(Mtr)-OH
Boc-Arg(Mtr)-OH

Sequence: Boc-Arg(Mtr)-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₂₁H₃₄N₄O₇S
MW: 486.59 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00043097
CAS Number: 102185-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR