Boc-N-Me-Phe-OH

Supplier: BACHEM AMERICAS INC

4001534.0001 4001534.0005
A-2075.0001BAEA 180.91 CAD
A-2075.0001BA A-2075.0005BA
Boc-N-Me-Phe-OH
Boc-N-Me-Phe-OH

Sequence: Boc-N-Me-Phe-OH


Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.
Short N-methyl-phenylalanine-rich peptides were shown to function as versatile blood-brain barrier shuttles.

Formula: C₁₅H₂₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD00038754
CAS Number: 37553-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR