Boc-N-methyl-L-isoleucine

Supplier: BACHEM AMERICAS INC

Synonyms: (2S,3S)-2-((tert-Butoxycarbonyl)(methyl)amino)-3-methylpentanoic acid, Boc-N-Me-Ile-OH

4001386.0001 4001386.0005
A-2055.0001BAEA 206.51 CAD
A-2055.0001BA A-2055.0005BA
Boc-N-methyl-L-isoleucine
Boc-N-methyl-L-isoleucine

Sequence: Boc-N-Me-Ile-OH

Formula: C₁₂H₂₃NO₄
MW: 245.319 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066105
CAS Number: 52498-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR