Boc-Cys(Mbzl)-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
N-α-t.-Boc-S-p-methylbenzyl-L-cysteine, Boc-Cys(pMeBzl)-OH
4010026.0025
4010026.0100
A-2030.0025BAEA
250.43
CAD
A-2030.0025BA
A-2030.0100BA
Boc-Cys(Mbzl)-OH
Boc-Cys(Mbzl)-OH
Sequence: Boc-Cys(Mbzl)-OH
The more acid-stable Mbzl group should be preferred over Mob when assembling large peptides by Boc-SPPS. Stable towards TFMSA/TFA, but readily removed by HF.
Formula:
C₁₆H₂₃NO₄S MW: 325.423 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00038517 CAS Number: 61925-77-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...