Boc-Aib-OH

Supplier: BACHEM AMERICAS INC

Synonyms: BOC-α-Methylalanine, N-Boc-alpha-methylalanine, 2-(Boc-amino)isobutyric Acid, Boc-Aib-OH

4010082.0025 4010082.0100
A-2015.0025BAEA 258.14 CAD
A-2015.0025BA A-2015.0100BA
Boc-Aib-OH
Boc-Aib-OH

Sequence: Boc-Aib-OH
Synonym(s): Boc-α-Me-Ala-OH

Formula: C₉H₁₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00042973
CAS Number: 30992-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR