Boc-Lys-OMe

Supplier: BACHEM AMERICAS INC

4002809.0001 4002809.0005
A-1925.0001BAEA 192.25 CAD
A-1925.0001BA A-1925.0005BA
Boc-Lys-OMe
Boc-Lys-OMe

Sequence: Boc-Lys-OMe


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
See also Z-Lys-ONp · HCl, M-1295.


This ester substrate is efficiently hydrolyzed by thrombin and especially trypsin.

Formula: C₁₂H₂₄N₂O₄
Storage Temperature: Freezer
MDL Number: MFCD00237564
CAS Number: 55757-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR