N-(ɑ)-Boc-L-lysine

Supplier: BACHEM AMERICAS INC

4000547.0005 4000547.0025
A-1920.0005BAEA 291.08 CAD
A-1920.0005BA A-1920.0025BA
N-(ɑ)-Boc-L-lysine
N-(alpha)-Boc-L-lysine

Sequence: Boc-Lys-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".


Educt for the synthesis of Boc- and Fmoc-ε-azido-norleucine.

Formula: C₁₁H₂₂N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00038203
CAS Number: 13734-28-6
EINECS: 237-303-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR